Learn More
Teknova 5% (w/v) Sodium Azide. 500mL, Sterile.
SDP
Supplier:  Teknova S0208
Encompass
Sodium azide is a versatile compound that has a wide range of applications in the laboratory. It can be used in dilute solutions as a preservative of biological samples and stock solutions, as well as an effective reagent for synthesis work.
Catalog No. 50-238-8091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.