Learn More
Teknova 2XYT Agar Plates with 2.0% Glucose and Carbenicillin-100.100mm, 20 Plates per Sleeve, Sterile. 4 Pack.
Supplier: Teknova Y43904PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin Disodium Salt