Learn More
Sino Biological Recombinant Mouse CNDP2 Protein (His Tag)
Supplier:  Sino Biological 50246M08B100
Encompass_Preferred
A DNA sequence encoding the mouse CNDP2 (NP_075638.2) (Met 1-Asn 475) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1629