Learn More
Sino Biological Recombinant Mouse Chk1 Protein (His & GST Tag)
Supplier:  Sino Biological 50248M20B50
Encompass_Preferred
A DNA sequence encoding the mouse CHEK1 (O35280-1) (Met1-Thr476) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-1634