Learn More
Sino Biological Recombinant Mouse PCSK9 Protein (mFc Tag)
Supplier:  Sino Biological 50251M05H20
Encompass_Preferred
A DNA sequence encoding the mouse PCSK9 (NP_705793.1) (Met1-Gln694) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-162-1643