Learn More
Bioworld HIV-1 p24 Core (Recombinant), 100 μg
Supplier:  Bioworld 220602111
Encompass_Preferred
Recombinant HIV-I p24 Source: E.coli Description : The protein contains the full-length sequence of HIV-I p24. Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein conc: 1mg/ml, 50 mM Tris pH 7.2 and 8M urea. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -200C. Stability: ReFive years frozen. One month in solution at room temperature. Specificity: Immunoreactive with all sera of HIV-I infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for early detection of HIV seroconvertors with minimal specificity problems. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-312