Learn More
ACROBiosystems Human EGF R 25tests
Supplier:  ACROBiosystems EGRHA2H825TESTS
Encompass_Preferred
APC-Labeled Human EGF R Protein His Tag (EGR-HA2H8) is produced via conjugation of APC to Human EGF R Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGF R Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).
Catalog No. 50-253-5118