Learn More
Trajan Scientific PEEKTUBE 1/16 ODX0.005 IDX10FT
Supplier: Trajan Scientific 130200500110F
Trajan Scientific and Medical PEEK Tubing, 1/16 in. O.D., 10 ft, Red, Diameter Inner: 0.005 in., Diameter Outer: 1/16 in., Overall Length: 10 ft., Maximum Pressure: 7000 psi (483 bar), Max. Operating Temperature: 100 deg.C, pH Range: 2 to 14