Learn More
Kyfora Bio Polyethylenimine Hydrochloride, Linear MW 160,000 (LPEI 160k) 2GR
Supplier:  Kyfora Bio 254392
Encompass_Preferred
L-PEI 160K is a highly-charged linear cationic polymer that readily binds highly anionic substrates. In research, LPEI 160k can be used to make composites for CO2 sensing. Key application(s): CO2 sensing
Catalog No. 50-255-9819