Learn More
Kyfora Bio Polyethylenimine, Linear, MW 2,500 (LPEI 2.5K) 2GR
Supplier:  Kyfora Bio 243132
Encompass
Linear polyethylenimine (L-PEI 2500) is a high-charge cationic polymer that readily binds highly anionic substrates. Low molecular weight linear PEIs, like L-PEI 2.5k, can be used for delivery of nucleic acids in vitro. Key application(s): Nucleic acid delivery
Catalog No. 50-255-9824