Learn More
Sino Biological Recombinant Mouse Protein C Protein (His Tag)
Supplier:  Sino Biological 50257M08H20
Encompass_Preferred
A DNA sequence encoding the mouse PROC (P33587) (Met1-Leu460) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1651