Learn More
Sino Biological Recombinant Mouse DR6/TNFRSF21 Protein (His Tag)
Supplier:  Sino Biological 50263M08H100
Encompass_Preferred
A DNA sequence encoding the mouse TNFRSF21 (NP_848704.1) extracellular domain (Met 1-His 349) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1658