Learn More
Sino Biological Recombinant Mouse FKBP12 Protein (His Tag)
Supplier:  Sino Biological 50265M07E100
Encompass_Preferred
A DNA sequence encoding the mouse FKBP1A (NP_032045.1) (Gly 2-Glu 108) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-162-1660