Learn More
Sino Biological Recombinant Mouse Dectin-2/CLEC6A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50267M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CLEC4N (NP_064385.1) extracellular domain (Ile 44-Leu 209) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1664