Learn More
ACROBiosystems CHO/Human STEAP1 Stable Cell Line Development Service
Supplier:  ACROBiosystems SCCHOATP1212VIALS
Encompass_Preferred
Six-transmembrane epithelial antigen of the prostate 1 (STEAP1) is an integral membrane protein that is highly up-regulated on the cell surface of several human cancers, making it a promising therapeutic target to manage these diseases. It shares sequence homology with three enzymes (STEAP2-STEAP4) that catalyze the NADPH-dependent reduction of iron(III). Taking into account its high specificity and overexpression in human cancers, STEAP1 is nowadays a promising candidate to be imposed as a therapeutic target.
Catalog No. 50-271-8940
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.