Learn More
PHCbi 11.5 cu.ft. Ref and 4.8 cu.ft. Frzr Pharmaceutical Refrigerator Freezer Combo
Supplier:  PHCbi MPRN450FHPA
Encompass_Preferred
PHCbi brand pharmaceutical refrigerator/freezer combos are engineered from the inside out to create, maintain and restore precise temperatures necessary for vaccine storage. Designed specifically for the storage of vaccines and other pharmaceuticals, our purpose-built refrigerator/freezer combos are designed for temperature uniformity and stability that is not commonly found with household or commercial cabinets. Separate refrigerator and freezer compartments provide a wider range of temperature storage points to accommodate a variety of vaccine product cold chain requirements.
Rigid polyurethane foamed-in-place
(3) Tempered glass, adjustable (Ref) | (2) PE coated wire (Frz)
2ºC~+14ºC (Refrigerator); -35ºC~-15ºC (Freezer)
Catalog No. 50-272-0040
Add to Cart
Request a Quote