Learn More
Sino Biological Recombinant Mouse ACP5 Protein (His Tag)
Supplier:  Sino Biological 50274M08H20
Encompass_Preferred
A DNA sequence encoding the pro form of mouse ACP5 (NP_031414.1) (Met 1-Pro 327) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1673