Learn More
Sino Biological Recombinant Mouse AGRP Protein (hFc Tag)
Supplier:  Sino Biological 50276M01H200
Encompass_Preferred
A DNA sequence encoding the mouse Agrp (NP_031453.1) (Ser82-Thr131) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1677