Learn More
Sino Biological Recombinant Mouse AGRP Protein (His Tag)
Supplier:  Sino Biological 50276M08H200
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse AGRP (P56473) (Met 1-Thr 131) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1681