Learn More
Sino Biological Recombinant Mouse Renin Protein (His Tag)
Supplier:  Sino Biological 50279M08H50
Encompass_Preferred
A DNA sequence encoding the pro form of mouse REN1 (NP_112469.1) (Met 1-Arg 402) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1685