Learn More
Sino Biological Recombinant Mouse IL-6R Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50280M08H50
Encompass_Preferred
A DNA sequence encoding the mouse IL6Rα (NP_034689.2) extracellular domain (Leu20-Glu357) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1687