Learn More
Sino Biological Recombinant Mouse LRPAP1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50281M08H100
Encompass_Preferred
A DNA sequence encoding the mature form of mouse LRPAP1 (NP_038615.2) extracellular domain (Gln 29-Leu 360) was fused with a signal peptide at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1688