Learn More
Sino Biological Recombinant Mouse CLEC4F Protein (ECD, hFc Tag)
Supplier:  Sino Biological 50282M01H50
Encompass_Preferred
A DNA sequence encoding the mouse Clec4f (NP_058031.2) (Arg70-Gly548) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1691