Learn More
Sino Biological Recombinant Mouse MARCO Protein (His Tag)
Supplier:  Sino Biological 50283M07H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse MARCO (NP_034896.1) (Gln 70-Ser 518) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1692