Learn More
Sino Biological Recombinant Mouse Cyclophilin A Protein (His Tag)
Supplier:  Sino Biological 50296M08E100
Encompass_Preferred
A DNA sequence encoding the mouse PPIA (NP_032933.1) (Met 1-Leu 164) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-162-1703