Learn More
Sino Biological Recombinant Mouse Angiopoietin-2/ANGPT2 Protein (His Tag)
Supplier:  Sino Biological 50298M07H50
Encompass_Preferred
A DNA sequence encoding the mouse ANGPT2 (NP_031452.2) (Lys275-Phe496) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-1708