Learn More
Sino Biological Recombinant Mouse Angiopoietin-1/ANGPT1 Protein (His Tag)
Supplier:  Sino Biological 50300M07H100
Encompass_Preferred
A DNA sequence encoding the mouse Angpt1 (NP_033770.2) (Arg277-Phe498) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1709