Learn More
Sino Biological Recombinant Mouse CLEC-2 Protein (hFc Tag)
Supplier:  Sino Biological 50306M01H100
Encompass_Preferred
A DNA sequence encoding the mouse Clec1b (NP_064369.1) (Met53-Leu229) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1718