Learn More
Sino Biological Recombinant Mouse Src Protein (His & GST Tag)
Supplier:  Sino Biological 50311M20B50
Encompass_Preferred
A DNA sequence encoding the mouse SRC (NP_001020566.111) (Met 1-Leu 535) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-1723