Learn More
Sino Biological Recombinant Mouse MEK1 Protein
Supplier:  Sino Biological 50312MNCB50
Encompass_Preferred
A DNA sequence encoding the mouse MAP2K1 (NP_032953.1) (Met1-Ile393) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-162-1727