Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 25/PK
Supplier: COLDER PRODUCTS COMPANY PMCD1702
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 3 2mm Buna-N Seal Overall Length 36 to 41 9 mm 1 45 to 1 65 in White Diameter Inner 0 125 in Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 0 81 in 20 5 mm