Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD1704
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 6 4mm Buna-N Seal Overall Length 41 to 45 7 mm 1 65 to 1 85 in White Diameter Inner 0 25 in Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 0 81 in 20 5 mm
Catalog No. 50-313-1293