Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polycarbonate Diameter Inner 6 4 mm 0 25 in Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple
Supplier:  COLDER PRODUCTS COMPANY MPC17004T03
Encompass_Preferred
Exact match for part number MPC17004T03
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polycarbonate Diameter Inner 6 4 mm 0 25 in Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple
Catalog No. 50-313-1303