Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Hose Barb Non-valved Coupling Insert Polysulfone w/Platinum-cured Silicone Seal Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Supplier:  COLDER PRODUCTS COMPANY MPC22004T39M
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-valved Coupling Insert Polysulfone w/Platinum-cured Silicone Seal Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 50-313-1319