Learn More
COLDER PRODUCTS COMPANY 1/4 HB ASEPTIQUIK G HIGH TEMP
Supplier:  COLDER PRODUCTS COMPANY AQG17004HT
Encompass_Preferred
Colder Products Company AseptiQuik G Connector Body High Temperature Hose Barb 1/4 in White with white pull tab Polycarbonate Silicone platinum-cured USP Class VI ADCF Maximum Pressure 60 psi Max Operating Temperature 104 deg F 40 deg C Min Operating Temperature 34 deg F 1 deg C
Catalog No. 50-313-1330