Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 10/PK
Supplier:  COLDER PRODUCTS COMPANY HFCD10812
Encompass_Preferred
CPC HFC Series High-Volume Quick-Disconnect Coupling Bodies Diameter Inner 0 5 in 9 5 mm Overall Length 2 9 in 73 6 mm Thread Size 0 5 in NPT Gray For Use With 12 7mm Includes Shut-off valves Polypropylene MNPT Pipe-thread Body Height 1 4 in 36 mm Maximum Pressure 60 psi
Catalog No. 50-313-1336