Learn More
COLDER PRODUCTS COMPANY QUICK DSCNNCT CPLNG BDY25PK
Supplier:  COLDER PRODUCTS COMPANY NS4D17006
Encompass_Preferred
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 60 45 mm 2 34 in Gray with Dark Gray Overmold For Use With 9 5mm Includes Shut-off valves Polypropylene Hose Barb Valved In-line Coupling Body Height 1 05 in 26 6 mm
Catalog No. 50-313-1354