Learn More
COLDER PRODUCTS COMPANY FEMALE COUPLING 1/4 25PK
Supplier:  COLDER PRODUCTS COMPANY NS4D10004
Encompass_Preferred
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 58 mm 2 31 in Gray with Dark Gray Overmold For Use With 6 4mm Includes Shut-off valves Polypropylene NPT Valved Coupling Body Height 1 05 in 26 6 mm Maximum Pressure 120 psi
Catalog No. 50-313-1361