Learn More
COLDER PRODUCTS COMPANY MALE INSRT 1/4 ID HB PNL 25PK
Supplier:  COLDER PRODUCTS COMPANY PLC42004
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 91 L x 0 94 in H (48 x 23 mm) Hose Barb Non-Valved Panel Mount Coupling Insert 0 25 in insert x 0 25 in hose barb White For Use With 6 4mm Includes Plastic connector Overall Length 1 91 in 48 mm
Catalog No. 50-313-1371