Learn More
COLDER PRODUCTS COMPANY CPC HFC35 Polysulfone Hose Barb Coupling Inserts 1/2 in hose barb valved in-line coupling insert
Supplier:  COLDER PRODUCTS COMPANY HFCD22835
Encompass_Preferred
CPC HFC35 Polysulfone Hose Barb Coupling Inserts 1/2 in hose barb valved in-line coupling insert
Catalog No. 50-313-1399