Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Sanitary Non-valved Coupling Body Polysulfone w/Platinum Cured Silicone Seal Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Supplier:  COLDER PRODUCTS COMPANY MPC3301239
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Sanitary Non-valved Coupling Body Polysulfone w/Platinum Cured Silicone Seal Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 50-313-1415