Learn More
COLDER PRODUCTS COMPANY 1/8 HB MICROCNX CONNECTOR
Supplier:  COLDER PRODUCTS COMPANY CNX17002HT
Encompass_Preferred
Colder Products Company MicroCNX Connector Hose Barb 1/8 in White with Teal Cover Polycarbonate USP Class VI ADCF Silicone platinum-cured USP Class VI ADCF Max Operating Temperature 104 deg F 40 deg C Height 0 93 in 23 6 mm Min Operating Temperature 34 deg F 1 deg C
Catalog No. 50-313-1420