Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMC2204
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 6 4mm Acetal Max Operating Temperature 82 deg C 180 deg F Min Operating Temperature -40 deg C -40 deg F White Diameter Inner 0 25 mm Overall Length 1 22 to 1 7 in 30 5 to 43 mm
Catalog No. 50-313-1435