Learn More
COLDER PRODUCTS COMPANY FEMALE BODY 3/8 ID HB 25/PK
Supplier:  COLDER PRODUCTS COMPANY PLC17006
Encompass_Preferred
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 8 L x 0 94 in H (45 7 x 23 mm) Hose Barb Non-Valved In-line Coupling Body 0 25 in flow coupler x 0 375 in hose barb White For Use With 9 5mm Includes Plastic connector Overall Length 1 8 in 45 7 mm
Catalog No. 50-313-1439