Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 10/PK
Supplier:  COLDER PRODUCTS COMPANY HFCD22812
Encompass_Preferred
CPC HFC Series High-Volume Quick-Disconnect Coupling Inserts With Shut-Off Valve Diameter Inner 0 5 in 9 5 mm Straight Hose-barb Insert Height 1 in 25 4 mm Overall Length 1 9 in 50 mm Gray For Use With 12 7mm Includes Shut-off valves Polypropylene Maximum Pressure 60 psi
Catalog No. 50-313-1449