Learn More
COLDER PRODUCTS COMPANY Insert AseptiQuik CPC 3/4 Sanitary Polycarbonate White w/blue pull tab Free floating Seal Silicone platinum-cured USP Class VI ADCF Gamma sterilized Temp Range 4 deg to 40 deg C Pressure Range Up to 60psi
Supplier:  COLDER PRODUCTS COMPANY AQC44012
Encompass_Preferred
Insert AseptiQuik CPC 3/4 Sanitary Polycarbonate White w/blue pull tab Free floating Seal Silicone platinum-cured USP Class VI ADCF Gamma sterilized Temp Range 4 deg to 40 deg C Pressure Range Up to 60psi
Catalog No. 50-313-1465