Learn More
COLDER PRODUCTS COMPANY Insert Coupling Colder Products Polysulfone USP Class VI ADCF 3/4 in hose barb Non valved Amber tint Free floating Seal Silicone platinum-cured USP Class VI ADCF Up to 60 psi 4 1 bar -40 to 300 deg F Gamma sterilized
Supplier: COLDER PRODUCTS COMPANY MPC44012T39M
Insert Coupling Colder Products Polysulfone USP Class VI ADCF 3/4 in hose barb Non valved Amber tint Free floating Seal Silicone platinum-cured USP Class VI ADCF Up to 60 psi 4 1 bar -40 to 300 deg F Gamma sterilized