Learn More
Sino Biological Recombinant Mouse Cortisol Binding Globulin/CBG Protein (His Tag)
Supplier:  Sino Biological 50314M08H100
Encompass_Preferred
A DNA sequence encoding the mouse SERPINA6 (Q06770) (Met 1-Ala 397) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1730