Learn More
Sino Biological Recombinant Mouse Nectin-2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50318M08H100
Encompass_Preferred
A DNA sequence encoding the mouse PVRL2 (NP_033016.3) extracellular domain (Met 1-Gly 351) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1741