Learn More
Sino Biological Recombinant Mouse TrkC Protein (His Tag)
Supplier:  Sino Biological 50320M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TrkC (NP_032772.3) (Met 1-Thr 429) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1745